Learn More
MgcRacGAP/RACGAP1 Rabbit anti-Human, Mouse, Rat, Clone: 10F7U6, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-15700-20UL
This item is not returnable.
View return policy
Description
MgcRacGAP/RACGAP1 Monoclonal antibody specifically detects MgcRacGAP/RACGAP1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| MgcRacGAP/RACGAP1 | |
| Monoclonal | |
| Unconjugated | |
| GTPase activating protein, HsCYK-4, ID-GAP, KIAA1478, Male germ cell RacGap, MGCRACGAP, MgcRacGAPrac GTPase-activating protein 1, Rac GTPase activating protein 1 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human MgcRacGAP/RACGAP1 (Q9H0H5). | |
| 20 μg | |
| Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
| 29127 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 10F7U6 | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |