Learn More
Novus Biologicals™ UBE2R2 Recombinant Protein Antigen
Product Code. 18354743 Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
Product Code. 18354743
Brand: Novus Biologicals™ NBP317955PEP
Recombinant Protein
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Antibody Competition
Formulation PBS and 1M Urea, pH 7.4.
Gene ID (Entrez) 54926
Molecular Weight (g/mol) M.W. Theoretical: 22 kDa
Quantity 100 μg
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Gene Alias CDC34BEC 6.3.2.19, E2-CDC34B, FLJ20419, MGC10481, UBC3BUbiquitin-protein ligase R2, Ubiquitin carrier protein R2, ubiquitin-conjugating enzyme E2 R2, Ubiquitin-conjugating enzyme E2-CDC34B, ubiquitin-conjugating enzyme E2R 2, ubiquitin-conjugating enzyme UBC3B
Gene Symbol UBE2R2
Research Category Cell Biology
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein UBE2R2