Learn More
Coiled-Coil Domain Containing 149 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-84710-0.1ML
This item is not returnable.
View return policy
Description
Coiled-Coil Domain Containing 149 Polyclonal specifically detects Coiled-Coil Domain Containing 149 in Human samples. It is validated for Western Blot.
Specifications
| Coiled-Coil Domain Containing 149 | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 91050 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| CCDC149, Coiled-Coil Domain-Containing Protein 149 | |
| The immunogen is a synthetic peptide directed towards the N-terminal region of Human Coiled-Coil Domain Containing 149 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido