Learn More
DUSP13 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-86621-0.1ML
Product Code. 18346901
  • £361.00 / 0.10mL
Description
DUSP13 Polyclonal specifically detects DUSP13 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DUSP13
Polyclonal
PBS, 2% Sucrose
Rabbit
Immunogen affinity purified
RUO
Primary
Human
Purified
Western Blot
Unconjugated
Branching-enzyme interacting DSP, dual specificity phosphatase 13, Dual specificity phosphatase SKRP4, dual specificity protein phosphatase 13, dual specificity protein phosphatase 13 isoform MDSP, dual-specificity phosphatase SKRP4, DUSP13A, DUSP13B, EC 3.1.3.16, EC 3.1.3.48, FLJ32450, MDSPbranching-enzyme interacting dual-specificity protein phosphatase, Muscle-restricted DSP, Testis- and skeletal-muscle-specific DSP, TMDPBEDPSKRP4
The immunogen for Anti-DUSP13 antibody is: synthetic peptide directed towards the N-terminal region of Human DUSP13
0.1 mL
Cell Cycle and Replication
51207
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.