Learn More
SLC22A14 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-84263-0.1ML
This item is not returnable.
View return policy
Description
SLC22A14 Polyclonal specifically detects SLC22A14 in Human samples. It is validated for Western Blot.
Specifications
| SLC22A14 | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 9389 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| MGC163415, OCTL2, OCTL4, ORCTL4, ORCTL-4, Organic cation transporter-like 4, organic cationic transporter-like 4, solute carrier family 22 (organic cation transporter), member 14, solute carrier family 22 member 14, solute carrier family 22, member 14 | |
| The immunogen is a synthetic peptide directed towards the N-terminal region of Human SLC22A14 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |