Learn More
OR8I2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-09829-100UL
This item is not returnable.
View return policy
Description
OR8I2 Polyclonal specifically detects OR8I2 in Human samples. It is validated for Western Blot.
Specifications
| OR8I2 | |
| Polyclonal | |
| Western Blot 1.0 ug/ml | |
| olfactory receptor 8I2, Olfactory receptor OR11-170, olfactory receptor, family 8, subfamily I, member 2, OR11-170 | |
| The immunogen is a synthetic peptide directed towards the C-terminal region of Human OR8I2 (NP_001003750). Peptide sequence YGSLIFTYLQPDNTSSLTQAQVASVFYTIVIPMLNPLIYSLRNKDVKNAL | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS buffer, 2% sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 120586 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |