Learn More
TBX20 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-86846
Product Code. 18334881
  • £362.00 / 100µL
Description
TBX20 Polyclonal specifically detects TBX20 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TBX20
Polyclonal
Unconjugated
PBS, 2% Sucrose
Rabbit
Affinity purified
RUO
57057
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
0.5 mg/ml
Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin
ASD4, T-box 20, T-box protein 20, T-box transcription factor TBX20
The immunogen is a synthetic peptide directed towards the N-terminal region of human TBX20