Learn More
BRN3B/POU4F2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-86585-0.1ML
This item is not returnable.
View return policy
Description
BRN3B/POU4F2 Polyclonal specifically detects BRN3B/POU4F2 in Human samples. It is validated for Western Blot.
Specifications
| BRN3B/POU4F2 | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| POU domain, class 4, transcription factor 2, BRN3.2, Brn-3b, BRN3B, POU class 4 homeobox 2 | |
| The immunogen is a synthetic peptide directed towards the N-terminal region of human BRN3B/POU4F2 | |
| 0.1 mL | |
| Neuroscience | |
| 5458 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |