Learn More
RB associated KRAB repressor Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-85609-0.1ML
This item is not returnable.
View return policy
Description
RB associated KRAB repressor Polyclonal specifically detects RB associated KRAB repressor in Human samples. It is validated for Western Blot.
Specifications
| RB associated KRAB repressor | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 57786 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| hRBaK, RB-associated KRAB repressor, RB-associated KRAB zinc finger, RB-associated KRAB zinc finger protein, ZNF769Zinc finger protein 769 | |
| The immunogen is a synthetic peptide directed towards the middle region of human RB associated KRAB repressor | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |