Learn More
RECK Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-10279-100UL
This item is not returnable.
View return policy
Description
RECK Polyclonal specifically detects RECK in Mouse samples. It is validated for Western Blot.
Specifications
| RECK | |
| Polyclonal | |
| Western Blot 1.0 ug/ml | |
| hRECK, membrane-anchored glycoprotein (metastasis and invasion), reversion-inducing-cysteine-rich protein with kazal motifs, ST15reversion-inducing cysteine-rich protein with Kazal motifs, suppression of tumorigenicity 15 (reversion-inducing-cysteine-rich protein withkazal motifs), suppression of tumorigenicity 5 (reversion-inducing-cysteine-rich protein withkazal motifs), Suppressor of tumorigenicity 15 protein | |
| The immunogen is a synthetic peptide directed towards the N terminal region of mouse RECK (NP_057887.2). Peptide sequence MNSSLPGVFKKSDGWVGLGCCELAIGLECRQACKQASSKNDISKVCRKEY | |
| 100 μg | |
| Cell Cycle and Replication | |
| 8434 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| PBS buffer, 2% sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |