Learn More
NFKBIL1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-84178-0.1ML
This item is not returnable.
View return policy
Description
NFKBIL1 Polyclonal specifically detects NFKBIL1 in Human samples. It is validated for Western Blot.
Specifications
| NFKBIL1 | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 4795 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| IkappaBL, I-kappa-B-like protein, IKBLikappaBL, Inhibitor of kappa B-like protein, LST1, NF-kappa-B inhibitor-like protein 1, nuclear factor of kappa light polypeptide gene enhancer in B-cellsinhibitor-like 1NFKBIL | |
| The immunogen is a synthetic peptide directed towards the N terminal region of human NFKBIL1 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |