Learn More
SALL1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-17307-25UL
 View more versions of this product
Product Code. 18318176
  • £385.00 / 25µL
Description
SALL1 Polyclonal antibody specifically detects SALL1 in Human samples. It is validated for Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SALL1
Polyclonal
Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL
HSAL1, sal (Drosophila)-like 1, Sal-1, SAL1, sal-like 1 (Drosophila), sal-like protein 1, Spalt-like transcription factor 1, Zinc finger protein 794, Zinc finger protein SALL1, Zinc finger protein Spalt-1, ZNF794TBS
This antibody was developed against Recombinant Protein corresponding to amino acids: ISESTSSMQALSPSNSTQEFHKSPSIEEKPQRAVPSEFANGLSPTPVNGGALDLTSSHAEK
25 μg
Primary
Human
Purified
Immunofluorescence
Unconjugated
PBS, pH 7.2, 40% glycerol
Rabbit
Affinity purified
RUO
6299
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.