Learn More
TOMM40L Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-83706-0.1ML
This item is not returnable.
View return policy
Description
TOMM40L Polyclonal specifically detects TOMM40L in Human samples. It is validated for Western Blot.
Specifications
| TOMM40L | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 84134 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| FLJ12770, mitochondrial import receptor subunit TOM40B, Protein TOMM40-like, TOMM40BRP11-297K8.10, translocase of outer mitochondrial membrane 40 homolog (yeast)-like, translocase of outer mitochondrial membrane 40 homolog-like, translocase of outer mitochondrial membrane 40 homolog-like (yeast) | |
| The immunogen is a synthetic peptide directed towards the middle region of human TOMM40L | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |