Learn More
EIF4E3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-12463
Product Code. 18313593
  • £409.00 / 100µg
Description
EIF4E3 Polyclonal antibody specifically detects EIF4E3 in Drosophila samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EIF4E3
Polyclonal
Western Blot 1:500, ELISA 1:10000
eIF4E type 3, eIF-4E type 3, eIF-4E3, eIF4E-3, eukaryotic translation initiation factor 4E family member 3, eukaryotic translation initiation factor 4E member 3, eukaryotic translation initiation factor 4E type 3, MGC39820, MGC86971
Synthetic peptide taken within amino acid region 1-90 on eIF4E3 protein.
100 μg
Primary
Drosophila
Purified
Western Blot, ELISA
Unconjugated
Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide
Rabbit
Affinity-purified
RUO
317649
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.