Learn More
ZNF496 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-10442-100UL
This item is not returnable.
View return policy
Description
ZNF496 Polyclonal specifically detects ZNF496 in Mouse samples. It is validated for Western Blot.
Specifications
| ZNF496 | |
| Polyclonal | |
| Western Blot 1.0 ug/ml | |
| zinc finger protein 232, ZSCAN11Zinc finger and SCAN domain-containing protein 11 | |
| The immunogen is a synthetic peptide corresponding to a region of Mouse (NP_766529). Peptide sequence GKIFRWRVNFIRHLRSRREQKPHKCSVCGELFSDSEDLDGHLETHEAQKP | |
| 100 μg | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS buffer, 2% sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 84838 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |