Learn More
AARSD1 Antibody (012), DyLight 680, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-90276FR
Product Code. 18303760
  • £480.00 / 0.10mL
Description
AARSD1 Monoclonal specifically detects AARSD1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
AARSD1
Monoclonal
DyLight 680
alanyl-tRNA editing protein Aarsd1, alanyl-tRNA synthetase domain containing 1, Alanyl-tRNA synthetase domain-containing protein 1, MGC2744
This antibody was obtained from a rabbit immunized with purified, recombinant Human AARSD1 (Q9BTE6-2; Thr176-Glu525).
0.1 mL
Primary
Human
Purified
Western Blot, ELISA, Immunofluorescence
12
50 mM Sodium Borate
Rabbit
Protein A purified
RUO
80755
Store at 4°C in the dark.