Learn More
Epstein Barr Virus Nuclear Antigen 6 Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
Brand:  Novus Biologicals NBP2-66598
Product Code. 18262944
  • £428.00 / 250µg
Description
Epstein Barr Virus Nuclear Antigen 6 Polyclonal specifically detects Epstein Barr Virus Nuclear Antigen 6 in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EpsteinBarrVirusNuclearAntigen6
Polyclonal
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Epstein Barr Virus Nuclear Antigen 3C
Sheep were immunized with recombinant EBNA-3C protein (Uniprot: P03204).
250 μg
Virus
Purified
Immunoprecipitation, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Unconjugated
Delipidated, heat inactivated, lyophilized stable whole serum with No Preservative
Sheep
Whole Antiserum
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.