Learn More
Novus Biologicals™ Fetuin A/AHSG Antibody Pair [HRP]
Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Fetuin A/AHSG.
Specifications
Specifications
| Assay Range | 14.06 pg/mL |
| Assay Sensitivity | 14.06-900 pg/mL |
| Assay | Sandwich ELISA |
| Conjugate | HRP |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | Antibody Pairs |
| Specificity | This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Fetuin A/AHSG. |
| Target Species | Human |
| For Use With (Application) | Sandwich ELISA |
| Gene Alias | A2HS, AHS, alpha-2-HS-glycoprotein, alpha-2-Z-globulin, Ba-alpha-2-glycoprotein, FETUAba-alpha-2-glycoprotein, fetuin-A, HSGA |
| Show More |