Learn More
Novus Biologicals™ NRBP2 Recombinant Protein Antigen
Product Code. 18248644 Shop All Bio Techne Products
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. 18248644
Brand: Novus Biologicals™ NBP258227PEP
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to NRBP2. The NRBP2 Recombinant Protein Antigen is derived from E. coli. The NRBP2 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
TRUSTED_SUSTAINABILITY
Specifications
Gene ID (Entrez) 340371
Purification Method >80% by SDS-PAGE and Coomassie blue staining
Common Name NRBP2 Recombinant Protein Antigen
Content And Storage Store at −20C. Avoid freeze-thaw cycles.
Formulation PBS and 1M Urea, pH 7.4.
For Use With (Application) Blocking/Neutralizing, Control
Gene Alias DKFZp434P086, MGC138699, nuclear receptor binding protein 2, nuclear receptor-binding protein 2, pp9320, Transformation-related gene 16 protein, transformation-related protein 16, TRG16, TRG-16
Gene Symbol NRBP2
Label Type Unlabeled
Product Type Recombinant Protein Antigen
Quantity 100 μL
Regulatory Status RUO
Source E.Coli
Specific Reactivity This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-49847. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml