Learn More
RBPSUHL Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-84239-0.1ML
This item is not returnable.
View return policy
Description
RBPSUHL Polyclonal specifically detects RBPSUHL in Human samples. It is validated for Western Blot.
Specifications
| RBPSUHL | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 11317 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| RBPL, RBP-L, RBPSUHL, recombination signal binding protein for immunoglobulin kappa J region-like, recombining binding protein suppressor of hairless (Drosophila)-like, recombining binding protein suppressor of hairless-like protein, SUH, SUHL, Transcription factor RBP-L | |
| The immunogen is a synthetic peptide directed towards the N terminal region of human RBPSUHL | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |