Learn More
Novus Biologicals™ ZNFN1A4 Recombinant Protein Antigen
Product Code. 18216252 Shop All Bio Techne Products
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. 18216252
Brand: Novus Biologicals™ NBP255305PEP
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human IKZF4. Source: E.coli Amino Acid Sequence: VNSGGYEKDVELVAHHSLEPGFGSSLAFVGAEHLRPLRLPPTNCISELTPVISSVYTQMQPLPGRLELPGSREAGEGPEDLADGGPLLYRPRGPLTDPGA The ZNFN1A4 Recombinant Protein Antigen is derived from E. coli. The ZNFN1A4 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
TRUSTED_SUSTAINABILITY
Specifications
Gene ID (Entrez) 64375
Purification Method >80% by SDS-PAGE and Coomassie blue staining
Common Name IKZF4 Recombinant Protein Antigen
Content And Storage Store at −20°C. Avoid freeze-thaw cycles
Formulation PBS and 1M Urea, pH 7.4
For Use With (Application) Blocking/Neutralizing, Control
Gene Alias N/A
Gene Symbol IKZF4
Label Type Unlabeled
Product Type Recombinant Protein Antigen
Quantity 100 μL
Regulatory Status RUO
Source E.coli
Specific Reactivity This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-50088. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml
Show More Show Less
For research use only.