Learn More
Mouse Cathepsin E Antibody, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody
Brand:  R&D Systems MAB1130
 View more versions of this product
Product Code. 18190921
  • £487.00 / 500µg
Description
Cathepsin E Monoclonal specifically detects Cathepsin E in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cathepsin E
Monoclonal
LYOPH
Western Blot 1 ug/mL
P70269
CTSE
Mouse myeloma cell line NS0-derived recombinant mouse Cathepsin E Gln19-Pro397 Accession # P70269
500 μg
Primary
Detects mouse Cathepsin E in direct ELISAs and Western blots. It recognizes both the pro and active forms of recombinant mouse (rm) Cathepsin E. In Western blots, no cross-reactivity with rmCathepsin B, C, H, L, or recombinant human Cathepsin O, S, or V is observed.
Mouse
Purified
Western Blot
186513
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CATE, cathepsin E, CTSE, EC 3.4.23, EC 3.4.23.34, erythrocyte membrane aspartic proteinase, slow-moving proteinase
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
1510
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2a