Learn More
FANCE Antibody (L5-P1E6*G5), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-50400F
Product Code. 18174348
  • £360.00 / 0.10mL
Description
FANCE Monoclonal specifically detects FANCE in Human samples. It is validated for ELISA, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FANCE
Monoclonal
FITC
PBS with 0.05% Sodium Azide
FANCE
Ovalbumin-conjugated synthetic peptide - KAALKHLGP
0.1 mL
Cancer, DNA Repair, Genes Sensitive to DNA Damaging Agents
2178
Store at 4C in the dark.
IgG1 κ
ELISA, Immunohistochemistry
L5-P1E6*G5
ELISA, Immunohistochemistry
FACEFAE, Fanconi anemia group E protein, Fanconi anemia, complementation group E, Protein FACE
Mouse
Protein A purified