Learn More
Human FLRT2 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB2877
This item is not returnable.
View return policy
Description
FLRT2 Monoclonal specifically detects FLRT2 in Human samples. It is validated for Western Blot.
Specifications
| FLRT2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| O43155 | |
| FLRT2 | |
| Mouse myeloma cell line NS0-derived recombinant human FLRT2 Cys36-Ser539 Accession # O43155 | |
| 100 μg | |
| Primary | |
| Detects human FLRT2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human (rh) FLRT1 or rhFLRT3. | |
| Human | |
| Purified |
| Western Blot | |
| 367205 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Fibronectin Leucine Rich Transmembrane Protein 2, Fibronectin-Like Domain-Containing Leucine-Rich Transmembrane Protein 2, KIAA0405, Leucine-Rich Repeat Transmembrane Protein FLRT2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 23768 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |