Learn More
PSP94/MSMB Antibody (OTI3D5) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73625
This item is not returnable.
View return policy
Description
PSP94/MSMB Monoclonal specifically detects PSP94/MSMB in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| Prostate Secretory Protein/PSP | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:200 - 1:1000 | |
| beta-microseminoprotein, IGBFProstate secreted seminal plasma protein, immunoglobulin binding factor, Immunoglobulin-binding factor, microseminoprotein, beta-, MSP, MSPB, PN44Prostate secretory protein of 94 amino acids, prostatic secretory protein 94, PRPS, PRSP, PSP, PSP57, PSP94HPC13, PSP-94Seminal plasma beta-inhibin | |
| Mouse | |
| 10.7 kDa | |
| 100 μg | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2b |
| Western Blot | |
| OTI3D5 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| MSMB | |
| Purified prostate specific antigen (MSMB) from human seminal fluid | |
| Affinity Purified | |
| Cancer | |
| 4477 | |
| Human | |
| Lyophilized |