Learn More
PTH Antibody (3H9 + PTH/1175) - C-terminus, N-terminal - IHC-Prediluted, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-48404
Product Code. 18161868
  • £373.00 / 7mL
Description
PTH Monoclonal specifically detects PTH in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PTH
Monoclonal
Unconjugated
10mM PBS and 0.05% BSA with 0.05% Sodium Azide
PTH
A synthetic peptide around aa 1-34 of human mature-PTH-polypeptide (exact sequence is proprietary) (3H9); A recombinant fragment around aa 32-115 of human mature PTH-polypeptide (exact sequence is proprietary) (Uniprot: P01270)
Protein A or G purified
RUO
Primary
Epitope of this monoclonal antibody maps in the C-terminus of PTH, a hormone produced by the parathyroid gland that regulates the concentration of calcium and phosphorus in extracellular fluid. This hormone elevates blood Ca2+ levels by dissolving the salts in bone and preventing their renal excretion. It is produced in the parathyroid gland as an 84 amino acid single chain polypeptide. It can also be secreted as N-terminal truncated fragments or C-terminal fragments after intracellular degradation, as in case of hypercalcemia. Defects in this gene are a cause of familial isolated hypoparathyroidism (FIH); also called autosomal dominant hypoparathyroidism or autosomal dominant hypocalcemia. FIH is characterized by hypocalcemia and hyperphosphatemia due to inadequate secretion of parathyroid hormone. Symptoms are seizures, tetany and cramps. FIH exist both as autosomal dominant and recessive forms of hypoparathyroidism.
Store at 4C.
IgG2b κ
Immunohistochemistry (Paraffin)
3H9 + PTH/1175
Immunohistochemistry-Paraffin
Parathormone, Parathyrin, parathyroid hormone, parathyroid hormone 1, PTH1
Mouse
9 kDa
7 mL
Apoptosis, Cancer
5741
Human
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications