Learn More
Novus Biologicals™ Recombinant Human MESDC1 His Protein
Product Code. 18147127 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
0.5 mg
Unit Size:
0.1mg
0.50mg
Product Code. 18147127
Brand: Novus Biologicals™ NBP2516700.5MG
Highly purified. Generating reliable and reproducible results.
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-362 of Human MESDC1 The Recombinant Human MESDC1 Protein is derived from E. coli. The Recombinant Human MESDC1 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY
Specifications
Concentration 0.5mg/mL
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0) containing 0.1M NaCl, 10% glycerol, 1mM DTT
Gene ID (Entrez) 59274
Molecular Weight (g/mol) TMW: 40.1kDa
Quantity 0.5 mg
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias Mesoderm Development Candidate 1
Common Name MESDC1
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Show More Show Less
For research use only and is not approved for use in humans or in clinical diagnosis