Learn More
GPR109B/HM74 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-24488SS
 View more versions of this product
Product Code. 18136225
  • £167.00 / 25µg
Description
GPR109B/HM74 Polyclonal specifically detects GPR109B/HM74 in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GPR109B/HM74
Polyclonal
Unconjugated
0.2 ml PBS and 0.2% Gelatin with 0.05% Sodium Azide
HCAR3
A portion of amino acids 1-50 of human GPR109B was used as the immunogen.
0.025 mg
Apoptosis, Cancer, GPCR, Tumor Suppressors
8843
Human, Primate
IgG
Western Blot, Immunohistochemistry (Paraffin)
0.5 mg/mL
Western Blot 0.5-2 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml
G protein-coupled receptor 109B, G-protein coupled receptor 109B, G-protein coupled receptor HM74, G-protein coupled receptor HM74B, GTP-binding protein, HCA3, HCAR3, HM74B, HM74Puma-g, Niacin receptor 2, NIACR2, Nicotinic acid receptor 2, PUMAG, putative chemokine receptor
Rabbit
Affinity Purified
RUO
Primary
The amino acid sequence used as immunogen is 100% homologous in human and chimpanzee, 86% in rhesus monkey and 73% homologous in bovine and human REG3G.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.