Learn More
Novus Biologicals™ Recombinant Human SAE2 His Protein
Product Code. 18106617 Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
Product Code. 18106617
Brand: Novus Biologicals™ NBP250574100UG
Highly purified. Generating reliable and reproducible results. Applications: SDS-Page
A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 1-640 of Human SAE2 The Recombinant Human SAE2 Protein is derived from E. coli. The Recombinant Human SAE2 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY
Specifications
Concentration 0.5mg/mL
For Use With (Application) SDS-PAGE
Formulation In PBS (pH 7.4) containing 10% glycerol
Gene ID (Entrez) 10054
Molecular Weight (g/mol) TMW: 72.3kDa
Quantity 100 μg
Source Baculovirus
Storage Requirements Store at 4°C short term. Aliquot and store at −70°C long term. Avoid freeze-thaw cycles.
Gene Alias Anthracycline-associated resistance ARX, ARX, EC 6.3.2, EC 6.3.2.-, FLJ13058, HRIHFB2115, SAE2SUMO-1 activating enzyme subunit 2, SUMO1 activating enzyme subunit 2, SUMO-activating enzyme subunit 2, UBA2, ubiquitin-activating enzyme E1 homolog, Ubiquitin-like 1-activating enzyme E1B, ubiquitin-like modifier activating enzyme 2, UBLE1B
Common Name SAE2
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >85%, by SDS-PAGE
Show More Show Less
For research use only and is not approved for use in humans or in clinical diagnosis