Learn More
Human/Mouse/Rat MLK4 alpha Antibody, R&D Systems™
Description
MLK4 alpha Polyclonal specifically detects MLK4 alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
| Antigen | MLK4 alpha |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL |
| Gene Accession No. | CAC84639 |
| Gene Alias | dJ862P8.3, KIAA1804, MAP3K21, MLK4 |
| Gene Symbols | KIAA1804 |
| Host Species | Rabbit |
| Show More |