Learn More
Human/Mouse Active Caspase-3 Antibody, R&D Systems™
Rabbit Polyclonal Antibody has been used in 155 publications
Brand: R&D Systems AF835
This item is not returnable.
View return policy
Description
Caspase-3 Polyclonal specifically detects Caspase-3 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry.
Specifications
| Caspase-3 | |
| Polyclonal | |
| Unconjugated | |
| P42574 | |
| CASP3 | |
| KLH-coupled human Caspase-3 synthetic peptide CRGTELDCGIETD Accession # P42574 | |
| 50 μg | |
| Primary | |
| Distinguishes apoptotic cells from non-apoptotic cells in immunohistochemistry. Detects the p17 subunit of Caspase-3, but does not detect the precursor form in Western blot. We recommend R&D Systems Affinity Purified Anti-Caspase-3 (Catalog # AF-605-NA) or Monoclonal Anti-Cleaved-Caspase-3 (Catalog # MAB835) for Western blot applications. | |
| Human, Mouse | |
| IgG |
| Immunohistochemistry, Immunocytochemistry | |
| LYOPH | |
| Immunohistochemistry 1-15 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| Apopain, apoptosis-related cysteine protease, CASP3, CASP-3, caspase 3, apoptosis-related cysteine peptidase, caspase-3, Caspase3, CPP32, CPP-32, CPP32B, CPP32SREBP cleavage activity 1, Cysteine protease CPP32, EC 3.4.22, EC 3.4.22.56, LICE-1, PARP cleavage protease, procaspase3, Protein Yama, SCA-1, Yama | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 836 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |