Learn More
Human Cornulin Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF3607
This item is not returnable.
View return policy
Description
Cornulin Polyclonal specifically detects Cornulin in Human samples. It is validated for Western Blot.
Specifications
| Cornulin | |
| Polyclonal | |
| Unconjugated | |
| Q9UBG3 | |
| CRNN | |
| E. coli-derived recombinant human Cornulin Met1-Ser140 Accession # Q9UBG3 | |
| 100 μg | |
| Primary | |
| Detects human Cornulin in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| 53 kDa squamous epithelial-induced stress protein, 58 kDa heat shock protein, C1orf10, chromosome 1 open reading frame 10, cornulin, CRNN, DRC1, Gm1015, PDRC1, PDRC1,53 kDa putative calcium-binding protein, SEP53, SEP53C1orf10, Squamous epithelial heat shock protein 53, Tumor-related protein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 49860 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |