Learn More
Tocris Bioscience™ MRS 2578
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
MRS 2578 is a selective antagonist of P2Y6 nucleotide receptors; IC50 values are 37 and 98 nM at human and rat P2Y6 receptors respectively. Displays no activity at P2Y1, P2Y2, P2Y4 and P2Y11 receptors (IC50 > 10 μM). Inhibits agonist-induced cardiomyocyte contraction and UDP-induced phagocytosis.
Specifications
Specifications
| Quantity | 50 mg |
| Product Type | MRS 2578 |
| Molecular Weight (g/mol) | 472.66 |
| Inhibitors | MRS 2578 |