Learn More
Alloy Steel, Cr, AISI E52100, G52986, ARMI∣MBH
Product Code. 17957514
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Product Code. 17957514
Brand: ARMI/MBH IARM49EG
ARMI∣MBH produces high-quality metal alloy certified reference materials and set up standards for Spark-OES, ICP, and XRF elemental analysis. The offering includes steel, aluminum, titanium, nickel, cobalt, copper, magnesium, and more.
Alloy Steel, Cr, AISI E52100, G52986, Ø38x19mm
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Oxygen/Arsenic/Niobium/Nitrogen/Cobalt/Phosphorus/Titanium/Tin/Molybdenum/Aluminium/Nickel/Vanadium/Copper/Silicon/Manganese/Carbon/Chromium
CAS 7439-89-6
Quantity 1 Ea.
Form Disc
Concentration or Composition (by Analyte or Components) Oxygen 0.002 %/Arsenic 0.0029 %/Niobium 0.003 %/Nitrogen 0.003 %/Cobalt 0.006 %/Phosphorus 0.006 %/Titanium 0.006 %/Tin 0.0065 %/Molybdenum 0.017 %/Aluminium 0.024 %/Nickel 0.043 %/Vanadium 0.066 %/Copper 0.076 %/Silicon 0.248 %/Manganese 0.364 %/Carbon 1.03 %/Chromium 1.43 %