Learn More
Invitrogen™ MALT1 Polyclonal Antibody
Goat Polyclonal Antibody
Brand:  Invitrogen™ PA5142766
Product Code. 17898242
  • £406.00 / 100µL
Description
Additional Information: This antibody is expected to recognize reported isoform 2 (XP_225927.5) only. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
MALT1/Paracaspase (MALT1) was independently identified as a member of the human paracaspase family and an interacting partner of B-cell lymphoma (Uren et al., 2000). MALT1 is a caspase-like protein that contains an N-terminal death domain, two Ig-like domains, and a C-terminal caspase-like domain. It binds to Bcl10 through its Ig-like domains and cooperates with Bcl10 to activate NF-kappaB.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MALT1
Polyclonal
Unconjugated
MALT1
A630046N12; caspase-like protein; D430033E09Rik; DKFZp434L132; IMD12; MALT lymphoma-associated translocation; Malt1; malt-1; MALT1 paracaspase; MALT1 protease; MLT; MLT1; mucosa associated lymphoid tissue lymphoma translocation gene 1; mucosa-associated lymphoid tissue lymphoma translocation protein 1; Mucosa-associated lymphoid tissue lymphoma translocation protein 1 homolog; paracaspase; paracaspase 1; paracaspase-1; PCASP1
Goat
Ammonium sulfate precipitation
RUO
240354, 307366
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Q2TBA3
MALT1
Peptide with sequence C-RPDNKEQTGQPLAK.
100 μg
Primary
Mouse, Rat