Learn More
BRAND™ Filter Tips Refill
GREENER_CHOICE
Product Code. 17887777 Shop All Brand Products
Click to view available options
Volume (Metric) Dispensing:
2 to 20 μL
5 to 100 μL
5 to 200 μL
50 to 1000 μL
50 to 1250 μL
Length (Metric):
50 mm
53 mm
70 mm
86 mm
102 mm
Unit Size:
Pack of 480
Pack of 960
Product Code. 17887777
Brand: BRAND™ 732636
Only null left
Add to basket
Add to basket
Cleanroom quality pipette tips designed for contamination-free analyses.
Pipette tips and filter tips from BRAND offer reliable quality for precise and demanding analyses. The raw materials are free from DiHEMDA und Oleamid. Tips are manufactured at BRAND under monitored cleanroom conditions and are automatically packaged. No parting agents or demolding aids are used. BRAND pipette tips are tested for BRAND pipettes and many pipette from other manufacturers.
BRAND pipette tips and filter tips as well as the corresponding TipBoxes are autoclavable at 121°C (2 bar) acc. DIN EN 285.
Sterile tips are manufactured under certified BIO-CERT™ quality. They are sterilized with beta radiation according to ISO 11 137 and the AAMI Guidelines.
Non-self-sealing filter tips from BRAND have a PE filter that is free from chemical additives. Permeability is controlled by the distinct combination of pore size and filter length, so that no aerosols can reach the pipette shaft.
BIO-CERT™ LIQUID HANDLING QUALITY: Free of human DNA, RNase, DNase, Pyrogens, PCR-Inhibitors and ATP
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) Dispensing 50 to 1250 μL
Filtered Filtered
Sterility Sterile
Color Colorless
Autoclavable Autoclavable
Length (Metric) 86 mm
Format Hanging Tip Rack
Tip Style Standard
Packaging Refill
Color Code Purple
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated No
Product Line Bio-Cert
Material Polypropylene
Fit for Purpose Certified Quality