Learn More
VEE GEE Plato Hydrometer with Thermometer
Click to view available options
Brix Range:
0 to 32° Plato
0 to 8.5° Plato
15.5 to 24° Plato
7.5 to 16° Plato
Unit Size:
Each
Description
- Calibration for use at 20°C/68°F
- Thermometer filled with SafetyBLUE liquid
Specifications
Specifications
| Type | Plato Hydrometer |
| Increments | 0.1° |
| Length (Metric) | 36 cm |
| Brix Range | 15.5 to 24° Plato |
| Material | Glass |
| Temperature Range (English) | 32°F to 122°F |
| Temperature Range (Metric) | 0°C to 50°C |
| Temperature of Standardization | 20°C (68°F) |