Learn More
CDK10 Polyclonal Antibody
CDK10 Polyclonal Antibody
Brand: Invitrogen PA5142464
This item is not returnable.
View return policy
Description
CDK10 Polyclonal Antibody for Western Blot
Specifications
| CDK10 | |
| Polyclonal | |
| Unconjugated | |
| CDK10 | |
| BC017131; CDC2-related protein kinase; Cdk10; Cell division protein kinase 10; cyclin dependent kinase 10; cyclin-dependent kinase (CDC2-like) 10; cyclin-dependent kinase 10; cyclin-dependent kinase related protein; PISSLRE; serine/threonine protein kinase PISSLRE; serine/threonine-protein kinase PISSLRE | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 8558 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q15131 | |
| CDK10 | |
| Peptide with sequence AEPDLECEQIRLK (aa 2-14). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |