Learn More
Chemtronics Foamtips™ 6 in. Foamtip Swab, Polypropylene Handle, 0.75 x 0.25 in. Head
Shop All Texwipe Products
Click to view available options
Length (English):
6 in.
Unit Size:
Pack of 1200
Description
Features and Benefits
- Precision cleaning swabs with polyurethane foam head
- Purest medical grade foam
- Traps surface particles while cleaning
- High solvent capacity; holds solvent well
- Fiberless construction does not generate loose fibers or particles
- No adhesives or binders are used in the construction
- Economical for high quantity applications
Applications
- Clean excess adhesives after gluing
- Clean micro-mechanical devices
- Remove Flux residues from printed circuit boards
- Apply small precise quantities of adhesive or lubricants
Specifications
Specifications
| Length (English) | 6 in. |
| Handle Material | Polypropylene |
| Product Type | Swab |
| Tip Material | Foam |
| Tip Type | Rounded |