Learn More
Eurofins DiscoverX LIM Kinase 1 Protein, Active
Product Code. 17380504
Encompass_Preferred
Click to view available options
Quantity:
10 μg
250 μg
Unit Size:
10µg
250µg
Product Code. 17380504
Brand: Eurofins DiscoverX 14656
Active, N-terminal 6His-tagged, recombinant, human LIM Kinase 1, amino acids 285-638, for use in Kinase Assays.
LIM domain kinases 1 and 2 (LIMK1 and LIMK2) play a critical role in stimulus-induced remodeling of the actin cytoskeleton by linking signals from the Rho family GTPases. LIMKs specifically phosphorylate and inactivate cofilin, an actin-binding protein that stimulates depolymerization and severance of actin filaments, thereby regulating actin cytoskeletal reorganization. Inhibition of LIMK1 may be useful in the treatment of tumors, especially invasive breast and prostate cancer cells.
GenBank NM_002314
Specifications
Formulation mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.
Molecular Weight (g/mol) 41.2 kDa
Quantity 10 μg
Species Human
Recombinant Recombinant
Protein Tag N-terminal 6His
Expression System Recombinant enzyme expressed in Sf21 insect cells
Protein Form Truncated
Purity or Quality Grade ≥67%
Protein LIMK1, active