Learn More
Intelsius™ PathoShield™
missing translation for 'orderingAttributeHoverText'
Size:
4
5
8
Dimensions (LxWxH):
191 x 145 x 22 mm
275 x 179 x 43 mm
291 x 191 x 102 mm
missing translation for 'unitSize'
Pack of 25
Pack of 50
Description
- Complete Category B shipping solution includes flexible 95kPa tested compliant secondary, absorbent, labels, and relevant UN markings
- Range of sizes suitable for shipping through postal and courier networks
- Suitable for a wide range of primaries including blood vials, swabs and sample containers
- Supplied flat packed for efficient storage and shipping prior to use
Specifications
Specifications
| Length (English) Exterior | 11.4 in. |
| Width (English) Exterior | 7.5 in. |
| Includes | Pouch |
| Size | 4 |
| Height (English) Exterior | 4.0 in. |
| Type | Sample Transport Packaging |
| Dimensions (L x W) | Pouch: 228 x 250 mm (8.9 x 9.8 in.) |
| Dimensions (LxWxH) | 291 x 191 x 102 mm |