Learn More
Kern Sohn Bench Scale ECE
Product Code. 17311573
Encompass_Preferred
Click to view available options
Capacity:
10 kg
20 kg
50 kg
Unit Size:
Each
Product Code. 17311573
Brand: Kern Sohn ECE 20K2N
Battery operation, compact, and lightweight bench scale with recessed grips on the underside.
  • High mobility: Battery operation, compact, lightweight construction, recessed grips on the underside and low weight make this balance ideal for use in several locations
  • Particularly large weighing plate
  • Very fast display: steady weight values within 2 s
  • Simple and convenient 2-key operation
Specifications
Capacity 20 kg
Dimensions (LxW) Platform 260 x 320 mm
Repeatability 10 g