Learn More
Invitrogen™ CD2AP Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114714
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CD2AP.
This gene encodes a scaffolding molecule that regulates the actin cytoskeleton. The protein directly interacts with filamentous actin and a variety of cell membrane proteins through multiple actin binding sites, SH3 domains, and a proline-rich region containing binding sites for SH3 domains. The cytoplasmic protein localizes to membrane ruffles, lipid rafts, and the leading edges of cells. It is implicated in dynamic actin remodeling and membrane trafficking that occurs during receptor endocytosis and cytokinesis. Haploinsufficiency of this gene is implicated in susceptibility to glomerular disease.
Specifications
| CD2AP | |
| Polyclonal | |
| Unconjugated | |
| CD2AP | |
| adapter protein CMS; AL024079; C78928; Cas ligand with multiple SH3 domains; Cas ligand with multiple Src homology 3 (SH3) domains; CD2 associated protein; Cd2ap; CD2-associated protein; CMS; mesenchyme to epithelium transition protein, SH3 domains; mesenchyme-to-epithelium transition protein with SH3 domains 1; Mets1; METS-1; SH3 domain-containing adapter protein; SH3 domain-containing adaptor protein METS-1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 12488, 23607, 316258 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| F1LRS8, Q9JLQ0, Q9Y5K6 | |
| CD2AP | |
| A synthesized peptide derived from human CD2AP(Accession Q9Y5K6), corresponding to amino acid residues T188-E238. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |