Learn More
Invitrogen™ Phospho-C/EBP beta (Thr235) Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5117238
Product Code. 17279733
  • £465.00 / 100µL
Description
Positive Samples: NIH/3T3 Immunogen sequence: PGTPS.
This gene belongs to the forkhead family of transcription factors which is characterized by a distinct DNA-binding forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in the development of mesenchymal tissues.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Phospho-C/EBP beta (Thr235)
Polyclonal
Unconjugated
Cebpb
AGP/EBP; C EBP; c/EBP beta; C/EBPb; C/EBPbeta; C/EBP-beta; C/EBP-related protein 2; CCAAT enhancer binding protein beta; CCAAT/enhancer binding protein (C/EBP), beta; CCAAT/enhancer binding protein beta; CCAAT/enhancer-binding protein beta; CEBPB; Crp2; Il6dbp; IL-6DBP; interleukin 6-dependent DNA-binding protein; interleukin-6-dependent-binding protein; LAP; LIP; Liver activator protein; Liver-enriched inhibitory protein; Liver-enriched transcriptional activator; Nfil6; Nf-il6; NF-M; nuclear factor NF-IL6; nuclear factor of interleukin 6; nuclear factor-IL6; nuclear protein Il6; OTTMUSP00000017503; PP9092; Sfb; SF-B; Silencer factor B; TCF5; TCF-5; transcription factor 5; transcription factor C/EBP beta
Rabbit
Affinity chromatography
RUO
1051, 12608
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
P17676, P28033
Cebpb
Synthetic peptide
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications