Learn More
Invitrogen™ PRDX3 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5116556
Product Code. 17279053
  • £455.00 / 100µL
Description
Positive Samples: U-87MG, HeLa, LO2, A-549, Mouse kidney Immunogen sequence: PAVTQHAPYF KGTAVVNGEF KDLSLDDFKG KYLVLFFYPL DFTFVCPTEI VAFSDKANEF HDVNCEVVAV SVDSHFSHLA WINTPRKNGG LGHMNIALLS DLTKQISRDY GVLLEGSGLA LRGLFIIDPN GVIKHLSVND LPVGRSVEET LRLVKAFQYV ETHGEVCPAN WTPDSPTIKP SPAASKEYFQ KVNQ.
Peroxiredoxin (Prx) is a growing peroxidase family, whose mammalian members have been known to connect with cell proliferation, differentiation, and apoptosis. Many isoforms (about 50 proteins), collected in accordance to the amino acid sequence homology, containing active site cysteine residue, and the thiol-specific antioxidant activity, distribute throughout all the kingdoms. Among them, mammalian Prx consists of 6 different members grouped into typical 2-Cys, atypical 2-Cys Prx, and 1-Cys Prx. Except Prx VI belonging to 1-Cys Prx subgroup, the other five 2-Cys Prx isotypes have the thioredoxin-dependent peroxidase (TPx) activity utilizing thioredoxin, thioredoxin reductase, and NADPH as a reducing system. Mammalian Prxs are 20-30 kilodalton in molecular size and vary in subcellular localization: Prx I, II, and VI in cytosol, Prx III in mitochondria, Prx IV in ER and secretion, Prx V showing complicated distribution including peroxisome, mitochondria and cytosol.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PRDX3
Polyclonal
Unconjugated
PRDX3
antioxidant protein; antioxidant protein 1; anti-oxidant protein 1; Aop1; AOP-1; AW822249; D0Tohi1; Ef2l; HBC189; Mer5; mitochondrial thioredoxin dependent peroxide reductase; mitochondrial Trx dependent peroxide reductase; perioredoxin-3; peroxiredoxin 3; peroxiredoxin III; Peroxiredoxin-3; Prdx3; PRO1748; Protein MER5; Protein MER5 homolog; Prx; Prx 3; PRx III; Prx3; PRX-3; Prx-III; SP22; SP-22; TDXM; thioredoxin-dependent peroxide reductase, mitochondrial
Rabbit
Affinity Chromatography
RUO
10935, 11757, 64371
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot, Immunocytochemistry
1.01 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
P20108, P30048, Q9Z0V6
PRDX3
Recombinant fusion protein containing a sequence corresponding to amino acids 63-256 of human Peroxiredoxin 3 (Peroxiredoxin 3 (PRDX3)) (NP_0067841).