Learn More
CD276 Mouse anti-Human, BUV496, Clone: 7-517, BD Biosciences™
Mouse Monoclonal Antibody
Brand:  BD Biosciences 749897
Product Code. 17261315
  • ONLINE EXCLUSIVE PRICE
    £356.00
    £341.54 / 50µg
    Save £14.46
    4% Off
Description
CD276 Monoclonal antibody specifically detects CD276 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD276
Monoclonal
Brilliant Ultraviolet 496
B7-H3, B7H3, 4Ig-B7-H3, B7RP-2
Human Monocyte-Derived Dendritic Cells
RUO
Human
IgG1 κ
Flow Cytometry
7-517
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse
50 μg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV496 under optimal conditions that minimize unconjugated dye and antibody.