Learn More
Invitrogen™ RECQL5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115832
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RECQL5.
RECQL5 gene ontology annotations related to this gene include cellular response to camptothecin; chromosome separation; DNA duplex unwinding; DNA recombination; DNA repair; DNA replication; double-strand break repair via homologous recombination; mitotic sister chromatid segregation; negative regulation of double-strand break repair via homologous recombination; negative regulation of mitotic cell cycle; negative regulation of transcription elongation from RNA polymerase II promoter; replication-born double-strand break repair via sister chromatid exchange; response to X-ray.
Specifications
| RECQL5 | |
| Polyclonal | |
| Unconjugated | |
| RECQL5 | |
| ATP-dependent DNA helicase Q5; DNA helicase, RecQ-like type 5; FLJ90603; LOW QUALITY PROTEIN: ATP-dependent DNA helicase Q5; RecQ helicase-like 5; RecQ like helicase 5; RecQ protein 5; RecQ protein-like 5; RecQ protein-like 5 like; RECQ5; Recq5b; RECQL5; Recql5b; RECQL5beta; recql5l | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 170472, 287834, 9400 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| D4ACP5, O94762, Q8VID5 | |
| RECQL5 | |
| A synthesized peptide derived from human RECQL5(Accession O94762), corresponding to amino acid residues E808-S858. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |