Learn More
CCR2 (CD192) Mouse anti-Human, BUV615, Clone: LS132.1D9 (also known as 1D9), BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 751045
This item is not returnable.
View return policy
Description
CCR2 (CD192) Monoclonal antibody specifically detects CCR2 (CD192) in Human samples. It is validated for Flow Cytometry
Specifications
| CCR2 (CD192) | |
| Monoclonal | |
| 0.2 mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. |
| Flow Cytometry | |
| LS132.1D9 (also known as 1D9) | |
| Brilliant Ultraviolet 615 | |
| CCR2; CD192; CKR2; CC-CKR-2; CMKBR2; MCP-1 receptor; MCP-1-R | |
| Human CCR2 Transfected Cell Line | |
| RUO | |
| Human | |
| IgG2a κ |