Learn More
Invitrogen™ NCK1 Recombinant Rabbit Monoclonal Antibody (4R1N9)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA535758
Product Code. 17176413
  • £438.00 / 100µL
Description
Immunogen sequence: PFSSSNDEEL NFEKGDVMDV IEKPENDPEW WKCRKINGMV GLVPKNYVTV MQNNPLTSGL EPSPPQCDYI RPSLTGKFAG NPWYYGKVTR HQAEMALNER G.
NCK1 is one of the signaling and transforming proteins containing Src homology 2 and 3 (SH2 and SH3) domains. It is located in the cytoplasm and is an adaptor protein involved in transducing signals from receptor tyrosine kinases to downstream signal recipients such as RAS. Alternatively spliced transcript variants encoding different isoforms have been found.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NCK1
Recombinant Monoclonal
0.38 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P16333, Q99M51
Nck1
A synthetic peptide corresponding to a sequence within amino acids 200-300 of human NCK11 (P16333).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
4R1N9
Unconjugated
Nck1
6330586M15Rik; Cytoplasmic protein NCK1; D230010O13Rik; melanoma NCK protein; MGC12668; Nck; NCK adaptor protein 1; NCK tyrosine kinase; NCK1; nck-1; NCKalpha; non-catalytic region of tyrosine kinase; non-catalytic region of tyrosine kinase adaptor protein 1; SH2/SH3 adaptor protein NCK-alpha
Rabbit
Affinity Chromatography
RUO
17973, 300955, 4690
-20°C, Avoid Freeze/Thaw Cycles