Learn More
RTDR1 Polyclonal Antibody
Brand: Thermo Scientific PA5113475
This item is not returnable.
View return policy
Description
Specifications
| RTDR1 | |
| Polyclonal | |
| Unconjugated | |
| RSPH14 | |
| 4933431K05Rik; MGC16968; Radial spoke head 14 homolog; radial spoke head homolog 14 (Chlamydomonas); rhabdoid tumor deletion region gene 1; rhabdoid tumor deletion region protein 1; Rsph14; Rtdr1 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 27156 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 4 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9UHP6 | |
| RSPH14 | |
| Recombinant Human Radial spoke head 14 homolog protein (1-230AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |